Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCCHC7
Entrez Gene ID
511821
Gene Name
zinc finger CCHC-type containing 7
Gene Aliases
-
Chromosome Location
Chr.8: 61209420 - 61541622 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046100.2 NP_001039565.1 4 - 5 926 129
GenBank mRNA BC112578.1 - 4 - 5 927 129

View Full Product Details