Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAGLN
Entrez Gene ID
513463
Gene Name
transgelin
Gene Aliases
-
Chromosome Location
Chr.15: 27889556 - 27894949 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046149.1 NP_001039614.1 1 - 2 64 62
GenBank mRNA BC105336.1 - 1 - 2 64 62

View Full Product Details