Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUV39H1
Entrez Gene ID
523047
Gene Name
suppressor of variegation 3-9 homolog 1
Gene Aliases
-
Chromosome Location
Chr.X: 86777328 - 86790238 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046264.2 NP_001039729.1 1 - 2 49 87
GenBank mRNA BC111156.1 - 1 - 2 50 87

View Full Product Details