Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCND1
Entrez Gene ID
524530
Gene Name
cyclin D1
Gene Aliases
-
Chromosome Location
Chr.29: 46889102 - 46898563 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046273.2 NP_001039738.1 2 - 3 572 86
GenBank mRNA BC112798.1 - 2 - 3 573 86

View Full Product Details