Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMTNL2
Entrez Gene ID
532143
Gene Name
smoothelin like 2
Gene Aliases
-
Chromosome Location
Chr.19: 25091270 - 25113046 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046320.1 NP_001039785.1 3 - 4 813 120
GenBank mRNA BC112730.1 - 3 - 4 813 120

View Full Product Details