Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HADH
Entrez Gene ID
532785
Gene Name
hydroxyacyl-CoA dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.6: 17208775 - 17252208 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046334.1 NP_001039799.1 2 - 3 367 66
GenBank mRNA BC105411.1 - 2 - 3 367 66

View Full Product Details