Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GBA
Entrez Gene ID
537087
Gene Name
glucosylceramidase beta
Gene Aliases
-
Chromosome Location
Chr.3: 15390177 - 15409497 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046421.2 NP_001039886.1 5 - 6 652 70
GenBank mRNA BC112823.1 - 5 - 6 653 70

View Full Product Details