Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERI3
Entrez Gene ID
540516
Gene Name
ERI1 exoribonuclease family member 3
Gene Aliases
PRNPIP
Chromosome Location
Chr.3: 101701172 - 101832068 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046479.2 NP_001039944.1 1 - 2 90 69
GenBank mRNA BC110159.1 - 1 - 2 91 69

View Full Product Details