Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A31
Entrez Gene ID
541168
Gene Name
solute carrier family 25 member 31
Gene Aliases
-
Chromosome Location
Chr.17: 29880313 - 29908490 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046500.2 NP_001039965.1 1 - 2 301 89
GenBank mRNA BC110266.1 - 1 - 2 303 89

View Full Product Details