Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSIG4
Entrez Gene ID
614262
Gene Name
V-set and immunoglobulin domain containing 4
Gene Aliases
-
Chromosome Location
Chr.X: 94489507 - 94560366 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046529.2 NP_001039994.1 3 - 4 550 79
GenBank mRNA BC112495.1 - 3 - 4 551 79

View Full Product Details