Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPACA1
Entrez Gene ID
614884
Gene Name
sperm acrosome associated 1
Gene Aliases
-
Chromosome Location
Chr.9: 61867831 - 61898286 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046545.1 NP_001040010.1 5 - 6 754 146
GenBank mRNA BC110228.1 - 5 - 6 754 146

View Full Product Details