Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC22C
Entrez Gene ID
614905
Gene Name
SEC22 homolog C, vesicle trafficking protein
Gene Aliases
-
Chromosome Location
Chr.22: 14697243 - 14723142 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046546.2 NP_001040011.1 1 - 2 168 70
GenBank mRNA BC110195.1 - 1 - 2 169 70

View Full Product Details