Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFU1
Entrez Gene ID
615964
Gene Name
NFU1 iron-sulfur cluster scaffold
Gene Aliases
-
Chromosome Location
Chr.11: 67778906 - 67807982 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046566.2 NP_001040031.1 4 - 5 363 156
GenBank mRNA BC105369.1 - 4 - 5 364 156

View Full Product Details