Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C25H16orf71
Entrez Gene ID
618674
Gene Name
chromosome 25 C16orf71 homolog
Gene Aliases
C16orf71
Chromosome Location
Chr.25: 3820604 - 3831178 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046621.2 NP_001040086.1 7 - 8 1067 89
GenBank mRNA BC112526.1 - 7 - 8 1068 89

View Full Product Details