Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM71D
Entrez Gene ID
618905
Gene Name
family with sequence similarity 71 member D
Gene Aliases
C10H14orf54, C14orf54
Chromosome Location
Chr.10: 79106907 - 79142204 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046625.1 NP_001040090.1 4 - 5 606 93
GenBank mRNA BC110256.1 - 4 - 5 606 93

View Full Product Details