Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC11C
Entrez Gene ID
504842
Gene Name
SEC11 homolog C, signal peptidase complex subunit
Gene Aliases
-
Chromosome Location
Chr.24: 58152254 - 58163466 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075175.2 NP_001068643.1 5 - 6 601 130
GenBank mRNA BC112783.1 - 5 - 6 602 130

View Full Product Details