Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC61B
Entrez Gene ID
506986
Gene Name
SEC61 translocon subunit beta
Gene Aliases
-
Chromosome Location
Chr.8: 64260374 - 64268676 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075292.2 NP_001068760.1 3 - 4 511 84
GenBank mRNA BC111652.1 - 3 - 4 512 84

View Full Product Details