Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC196
Entrez Gene ID
513215
Gene Name
coiled-coil domain containing 196
Gene Aliases
C10H14orf53, C14orf53
Chromosome Location
Chr.10: 78535551 - 78560301 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075604.2 NP_001069072.2 10 - 11 808 80
NR_045536.1 10 - 11 968 80
GenBank mRNA BC110259.1 - 10 - 11 969 80

View Full Product Details