Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC19
Entrez Gene ID
514466
Gene Name
zinc finger DHHC-type palmitoyltransferase 19
Gene Aliases
-
Chromosome Location
Chr.1: 70789424 - 70802153 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075671.2 NP_001069139.1 2 - 3 351 71
GenBank mRNA BC112567.1 - 2 - 3 353 71

View Full Product Details