Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC63
Entrez Gene ID
518001
Gene Name
coiled-coil domain containing 63
Gene Aliases
-
Chromosome Location
Chr.17: 54667572 - 54705548 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075763.2 NP_001069231.1 10 - 11 1537 80
GenBank mRNA BC111238.1 - 10 - 11 1539 80

View Full Product Details