Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPS2
Entrez Gene ID
518494
Gene Name
G protein pathway suppressor 2
Gene Aliases
-
Chromosome Location
Chr.19: 27020131 - 27023339 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075774.2 NP_001069242.1 9 - 10 894 98
GenBank mRNA BC109646.1 - 9 - 10 895 98

View Full Product Details