Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SBSPON
Entrez Gene ID
525332
Gene Name
somatomedin B and thrombospondin type 1 domain containing
Gene Aliases
C14H8orf84, RPESP
Chromosome Location
Chr.14: 36429760 - 36467141 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075869.2 NP_001069337.1 1 - 2 270 63
GenBank mRNA BC109766.1 - 1 - 2 273 63

View Full Product Details