Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNL4A
Entrez Gene ID
535457
Gene Name
thioredoxin like 4A
Gene Aliases
-
Chromosome Location
Chr.24: 539078 - 549835 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076060.1 NP_001069528.1 3 - 4 412 70
GenBank mRNA BC109892.1 - 3 - 4 412 70

View Full Product Details