Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZD9
Entrez Gene ID
537386
Gene Name
PDZ domain containing 9
Gene Aliases
C16orf65, C25H16orf65
Chromosome Location
Chr.25: 19669081 - 19693882 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076094.1 NP_001069562.1 3 - 4 481 97
GenBank mRNA BC109887.1 - 3 - 4 481 97

View Full Product Details