Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRAP1
Entrez Gene ID
540345
Gene Name
DR1 associated protein 1
Gene Aliases
-
Chromosome Location
Chr.29: 44028245 - 44030520 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076214.1 NP_001069682.1 2 - 3 210 83
GenBank mRNA BC110127.1 - 2 - 3 210 83

View Full Product Details