Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPRT
Entrez Gene ID
540851
Gene Name
uracil phosphoribosyltransferase homolog
Gene Aliases
-
Chromosome Location
Chr.X: 75911721 - 75941962 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076245.2 NP_001069713.1 5 - 6 767 142
GenBank mRNA BC109679.1 - 5 - 6 770 142

View Full Product Details