Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDCA5
Entrez Gene ID
613318
Gene Name
cell division cycle associated 5
Gene Aliases
-
Chromosome Location
Chr.29: 43280453 - 43297228 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076270.2 NP_001069738.1 2 - 3 218 84
GenBank mRNA BC110243.1 - 2 - 3 219 84

View Full Product Details