Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB8OS
Entrez Gene ID
613795
Gene Name
zinc finger and BTB domain containing 8 opposite strand
Gene Aliases
-
Chromosome Location
Chr.2: 120986507 - 121002878 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076288.2 NP_001069756.1 4 - 5 763 141
GenBank mRNA BC110257.1 - 4 - 5 764 141

View Full Product Details