Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YEATS4
Entrez Gene ID
615330
Gene Name
YEATS domain containing 4
Gene Aliases
-
Chromosome Location
Chr.5: 44086259 - 44110850 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076365.2 NP_001069833.1 4 - 5 600 127
GenBank mRNA BC109626.1 - 4 - 5 602 127

View Full Product Details