Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC39A11
Entrez Gene ID
615681
Gene Name
solute carrier family 39 member 11
Gene Aliases
-
Chromosome Location
Chr.19: 58126981 - 58503178 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076390.1 NP_001069858.1 8 - 9 842 140
GenBank mRNA BC110274.1 - 8 - 9 842 140

View Full Product Details