Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC12B
Entrez Gene ID
616085
Gene Name
C-type lectin domain family 12 member B
Gene Aliases
-
Chromosome Location
Chr.5: 99926493 - 99932836 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076408.2 NP_001069876.1 5 - 6 689 79
GenBank mRNA BC111146.1 - 5 - 6 690 79

View Full Product Details