Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UQCC1
Entrez Gene ID
618503
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 1
Gene Aliases
C20orf44, UQCC
Chromosome Location
Chr.13: 64573897 - 64669138 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076508.2 NP_001069976.1 2 - 3 351 66
GenBank mRNA BC110232.1 - 2 - 3 352 66

View Full Product Details