Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASRGL1
Entrez Gene ID
767970
Gene Name
asparaginase and isoaspartyl peptidase 1
Gene Aliases
-
Chromosome Location
Chr.29: 40756509 - 40775200 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077035.2 NP_001070503.1 1 - 2 110 118
GenBank mRNA BC109621.1 - 1 - 2 111 118

View Full Product Details