Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H12orf29
Entrez Gene ID
767986
Gene Name
chromosome 5 C12orf29 homolog
Gene Aliases
C12orf29
Chromosome Location
Chr.5: 17826835 - 17836741 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077047.2 NP_001070515.1 5 - 6 772 96
GenBank mRNA BC105491.1 - 5 - 6 773 96

View Full Product Details