Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSX5
Entrez Gene ID
767992
Gene Name
synovial sarcoma, X breakpoint 5
Gene Aliases
-
Chromosome Location
Chr.X: 86550751 - 86564476 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077053.2 NP_001070521.1 6 - 7 543 97
GenBank mRNA BC109709.1 - 6 - 7 544 97

View Full Product Details