Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COG8
Entrez Gene ID
768007
Gene Name
component of oligomeric golgi complex 8
Gene Aliases
-
Chromosome Location
Chr.18: 36474032 - 36480940 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077067.2 NP_001070535.1 4 - 5 1623 94
GenBank mRNA BC110177.1 - 4 - 5 1624 94

View Full Product Details