Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TERB2
Entrez Gene ID
768019
Gene Name
telomere repeat binding bouquet formation protein 2
Gene Aliases
C10H15orf43, C15orf43
Chromosome Location
Chr.10: 103201542 - 103245521 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077078.2 NP_001070546.1 5 - 6 461 105
GenBank mRNA BC111624.1 - 5 - 6 462 105

View Full Product Details