Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8H8orf74
Entrez Gene ID
768069
Gene Name
chromosome 8 C8orf74 homolog
Gene Aliases
C8orf74
Chromosome Location
Chr.8: 8587751 - 8618693 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077126.1 NP_001070594.1 2 - 3 264 70
GenBank mRNA BC111241.1 - 2 - 3 264 70

View Full Product Details