Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPY30
Entrez Gene ID
513162
Gene Name
dpy-30 histone methyltransferase complex regulatory subunit
Gene Aliases
-
Chromosome Location
Chr.11: 14665875 - 14678722 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077916.2 NP_001071384.1 1 - 2 67 62
GenBank mRNA BC111634.1 - 1 - 2 70 62

View Full Product Details