Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A5
Entrez Gene ID
777599
Gene Name
membrane spanning 4-domains A5
Gene Aliases
-
Chromosome Location
Chr.15: 83251372 - 83278961 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078146.2 NP_001071614.1 2 - 3 379 133
GenBank mRNA BC112628.1 - 2 - 3 380 133

View Full Product Details