Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMGT1
Entrez Gene ID
617557
Gene Name
membrane magnesium transporter 1
Gene Aliases
TMEM32
Chromosome Location
Chr.Unknown: 8925411 - 8937711 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079630.1 NP_001073098.1 3 - 4 531 145
GenBank mRNA BC109649.1 - 3 - 4 531 145

View Full Product Details