Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC46
Entrez Gene ID
780880
Gene Name
leucine rich repeat containing 46
Gene Aliases
-
Chromosome Location
Chr.19: 38627367 - 38631918 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079799.1 NP_001073267.1 4 - 5 559 94
GenBank mRNA BC112534.1 - 4 - 5 559 94

View Full Product Details