Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STAT5B
Entrez Gene ID
282376
Gene Name
signal transducer and activator of transcription 5B
Gene Aliases
-
Chromosome Location
Chr.19: 42319172 - 42384926 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174617.4 NP_777042.2 18 - 19 2322 110
GenBank mRNA AJ237933.1 - 17 - 18 2239 110
BC112568.1 - 18 - 19 2323 110

View Full Product Details