Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAT5B
Entrez Gene ID
282376
Gene Name
signal transducer and activator of transcription 5B
Gene Aliases
-
Chromosome Location
Chr.19: 42319172 - 42384926 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174617.4 NP_777042.2 2 - 3 215 88
GenBank mRNA AJ237933.1 - 1 - 2 132 88
BC112568.1 - 2 - 3 216 88

View Full Product Details