Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD53
Entrez Gene ID
505040
Gene Name
CD53 molecule
Gene Aliases
-
Chromosome Location
Chr.3: 32494128 - 32533026 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034232.2 NP_001029404.1 1 - 2 83 116
GenBank mRNA BC113322.1 - 1 - 2 83 116

View Full Product Details