Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPEF1
Entrez Gene ID
512410
Gene Name
sperm flagellar 1
Gene Aliases
C20orf28
Chromosome Location
Chr.13: 51421440 - 51425986 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034422.2 NP_001029594.1 3 - 4 545 85
GenBank mRNA BC118201.1 - 3 - 4 545 85
BT021696.1 - 3 - 4 526 85

View Full Product Details