Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TANGO2
Entrez Gene ID
518209
Gene Name
transport and golgi organization 2 homolog
Gene Aliases
C17H22orf25, C22orf25
Chromosome Location
Chr.17: 72949358 - 72967481 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035328.2 NP_001030405.2 5 - 6 491 105
GenBank mRNA BC113297.1 - 5 - 6 491 105

View Full Product Details