Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIMP4
Entrez Gene ID
317694
Gene Name
TIMP metallopeptidase inhibitor 4
Gene Aliases
-
Chromosome Location
Chr.22: 56946000 - 56952875 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045871.2 NP_001039336.1 2 - 3 286 117
GenBank mRNA BC112857.1 - 2 - 3 287 117

View Full Product Details