Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A19
Entrez Gene ID
504418
Gene Name
solute carrier family 25 member 19
Gene Aliases
-
Chromosome Location
Chr.19: 56205905 - 56217714 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045887.1 NP_001039352.1 1 - 2 28 94
GenBank mRNA BC114115.1 - 1 - 2 28 94

View Full Product Details