Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM256
Entrez Gene ID
507083
Gene Name
transmembrane protein 256
Gene Aliases
C17orf61, C19H17orf61
Chromosome Location
Chr.19: 27081977 - 27083126 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045957.1 NP_001039422.1 2 - 3 147 74
GenBank mRNA BC112791.1 - 2 - 3 147 74

View Full Product Details